Learn More
Cayman Chemical ANTIBODY O12-D3-I3 10mg
SDP
Catalog No. 502944228
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.39); has been used in the generation of LNPs for siRNA delivery in vitro and in vivo; LNPs containing O12-D3-I3 and encapsulating ferrocene and siRNA targeting Nrf2 or system xc- cystine/glutamate transporter decrease viability of HepG2 cells; LNPs containing O12-D3-I3 and an mRNA reporter selectively accumulate in the tumor and liver over the lungs, kidneys, spleen, and heart in mice; LNPs containing O12-D3-I3 and encapsulating ferrocene and siRNA targeting Nrf2 or system xc- cystine/glutamate transporter reduce tumor growth and volume in an MCF-7 breast cancer mouse xenograft model; LNPs containing O12-D3-I3 and encapsulating siRNA targeting Factor VII decrease hepatocyte Factor VII mRNA levels in mice
Catalog No. 50-294-4228 Supplier Cayman Chemical Supplier No. 4138510 MG
An ionizable cationic lipid (pKa = 6.39); has been used in the generation of LNPs for siRNA delivery in vitro and in vivo; LNPs containing O12-D3-I3 and encapsulating ferrocene and siRNA targeting Nrf2 or system xc- cystine/glutamate transporter decrease viability of HepG2 cells; LNPs containing O12-D3-I3 and an mRNA reporter selectively accumulate in the tumor and liver over the lungs, kidneys, spleen, and heart in mice; LNPs containing O12-D3-I3 and encapsulating ferrocene and siRNA targeting Nrf2 or system xc- cystine/glutamate transporter reduce tumor growth and volume in an MCF-7 breast cancer mouse xenograft model; LNPs containing O12-D3-I3 and encapsulating siRNA targeting Factor VII decrease hepatocyte Factor VII mRNA levels in mice