Learn More
Cayman Chemical OC2-K3-E10, Purity: ≥95%, CAS: 2933216-12-5, 50 mg
SDP
Catalog No. 502916255
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of lncRNA in mice and for the delivery of CRISPR complementary sgRNA and Cas9 mRNA for genome editing in transgenic mice; LNPs containing OC2-K3-E10 and encapsulating either GAPLINC or DRAIR lncRNA reduce LPS-induced increases in serum IL-1β or TNF-α and IL-6 levels, respectively, in mice; OC2-K3-E10-containing LNPs encapsulating ψ-modified MIST reduce LPS-induced increases in TNF-α levels in mice
Catalog No. 50-291-6255 Supplier Cayman Chemical Supplier No. 3824350 MG
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of lncRNA in mice and for the delivery of CRISPR complementary sgRNA and Cas9 mRNA for genome editing in transgenic mice; LNPs containing OC2-K3-E10 and encapsulating either GAPLINC or DRAIR lncRNA reduce LPS-induced increases in serum IL-1β or TNF-α and IL-6 levels, respectively, in mice; OC2-K3-E10-containing LNPs encapsulating ψ-modified MIST reduce LPS-induced increases in TNF-α levels in mice