Learn More
Cayman Chemical Odanacatib, Purity: ≥98%, CAS: 603139-19-1, 10 mg
SDP
Catalog No. 502985941
Encompass_Preferred
A potent, selective, and neutral inhibitor of cathepsin K (IC50s = 0.2 and 1 nM for human and rabbit enzymes, respectively)
Catalog No. 50-298-5941 Supplier Cayman Chemical Supplier No. 2146610 MG
A potent, selective, and neutral inhibitor of cathepsin K (IC50s = 0.2 and 1 nM for human and rabbit enzymes, respectively)