Learn More
Cayman Chemical OGG1-IN-O8, Purity: ≥98%, CAS: 350997-39-6, 10 mg
SDP
Catalog No. 502992316
Encompass_Preferred
An OGG1 inhibitor (IC50 = 0.35 µM); selective for OGG1 over NEIL1, NTH1, and Fpg at 50 µM
Catalog No. 50-299-2316 Supplier Cayman Chemical Supplier No. 2976910 MG
An OGG1 inhibitor (IC50 = 0.35 µM); selective for OGG1 over NEIL1, NTH1, and Fpg at 50 µM