Learn More
Cayman Chemical OH-Chol, Purity: ≥95%, CAS: 191173-82-7, 5 mg
SDP
Catalog No. 502934812
Encompass_Preferred
A cationic cholesterol derivative; has been used in lipoplexes with DOPE for siRNA delivery and gene silencing in MCF-7 cells as well as in mice via intravenous injection, resulting in lipoplex accumulation in the liver; has been used in cationic nanoparticles in combination with Tween 80 to transfect pDNA and siRNA into PC3 mouse xenografts via intratumoral injection; has been used in cationic nanoparticles in combination with Tween 80 and folate-PEG2000-DSPE in a KB mouse xenograft model for intratumoral gene delivery
Catalog No. 50-293-4812 Supplier Cayman Chemical Supplier No. 265865 MG
A cationic cholesterol derivative; has been used in lipoplexes with DOPE for siRNA delivery and gene silencing in MCF-7 cells as well as in mice via intravenous injection, resulting in lipoplex accumulation in the liver; has been used in cationic nanoparticles in combination with Tween 80 to transfect pDNA and siRNA into PC3 mouse xenografts via intratumoral injection; has been used in cationic nanoparticles in combination with Tween 80 and folate-PEG2000-DSPE in a KB mouse xenograft model for intratumoral gene delivery