Learn More
Cayman Chemical Ospemifene, Purity: ≥98%, CAS: 128607-22-7, 250 mg
SDP
Catalog No. 502994976
Encompass_Preferred
A selective estrogen receptor modulator; binds to ERα and ERβ (Kis = 380 and 410 nM, respectively); increases vaginal weight and vaginal epithelial height (ED50s = 0.48 and 0.39 mg/kg per day, respectively) in an ovariectomized rat model of menopause; increases progesterone receptor protein expression in vaginal epithelium and stroma and inhibits 17β-estradiol-induced, estrogen response element-mediated transactivation in a reporter assay using MCF-7 cells
Catalog No. 50-299-4976 Supplier Cayman Chemical Supplier No. 23755250 MG
A selective estrogen receptor modulator; binds to ERα and ERβ (Kis = 380 and 410 nM, respectively); increases vaginal weight and vaginal epithelial height (ED50s = 0.48 and 0.39 mg/kg per day, respectively) in an ovariectomized rat model of menopause; increases progesterone receptor protein expression in vaginal epithelium and stroma and inhibits 17β-estradiol-induced, estrogen response element-mediated transactivation in a reporter assay using MCF-7 cells