Learn More
Cayman Chemical p,p'-DDE, Purity: ≥98%, CAS: 72-55-9, 100 mg
SDP
Catalog No. 502935694
Encompass_Preferred
A metabolite and degradation product of the pesticide DDT; accumulates in various fish and sediment surrounding a DDT manufacturing plant; inhibits estrogen binding to rtERs (IC50 = 8 µM); induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; decreases ATP levels, the proportion of sperm with high mitochondrial membrane potential, and motility of human sperm in vitro from 50-100 µM
Catalog No. 50-293-5694 Supplier Cayman Chemical Supplier No. 24241100 MG
A metabolite and degradation product of the pesticide DDT; accumulates in various fish and sediment surrounding a DDT manufacturing plant; inhibits estrogen binding to rtERs (IC50 = 8 µM); induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; induces concentration-dependent secretion of estradiol by granulosa and theca cell co-cultures isolated from porcine ovarian follicles; decreases ATP levels, the proportion of sperm with high mitochondrial membrane potential, and motility of human sperm in vitro from 50-100 µM