Learn More
Cayman Chemical PAP-1, Purity: ≥98%, CAS: 870653-45-5, 5 mg
SDP
Catalog No. 502991211
Encompass_Preferred
An inhibitor of Kv1.3 channels (EC50 = 2 nM at -80 mV in cells overexpressing the human channel); selective for Kv1.3 over a panel of 22 ion channels (EC50s = 45-15,000 nM for all); inhibits anti-CD3 antibody-induced proliferation of isolated human CCR7- effector memory T cells; inhibits ovalbumin-induced delayed-type hypersensitivity in rats at 3 mg/kg
Catalog No. 50-299-1211 Supplier Cayman Chemical Supplier No. 301055 MG
An inhibitor of Kv1.3 channels (EC50 = 2 nM at -80 mV in cells overexpressing the human channel); selective for Kv1.3 over a panel of 22 ion channels (EC50s = 45-15,000 nM for all); inhibits anti-CD3 antibody-induced proliferation of isolated human CCR7- effector memory T cells; inhibits ovalbumin-induced delayed-type hypersensitivity in rats at 3 mg/kg