Learn More
Cayman Chemical Paquinimod, Purity: ≥98%, CAS: 248282-01-1, 10 mg
SDP
Catalog No. 502940380
Encompass_Preferred
An immunomodulator; reduces splenic counts of Ly6ChiCD115+ inflammatory monocytes and CD11b+CD11c+ dendritic cells in a mouse model of EAE; reduces hepatic fibrosis and accumulation of inflammatory monocytes and neutrophils in the N-IF mouse model of liver fibrosis at 25 mg/kg per day; delays disease onset and decreases insulitis in NOD mice; reduces skin thickness, the number of myofibroblasts, and autoantibody production in the tight skin 1 mouse model of systemic sclerosis at 5 and 25 mg/kg
Catalog No. 50-294-0380 Supplier Cayman Chemical Supplier No. 2844310 MG
An immunomodulator; reduces splenic counts of Ly6ChiCD115+ inflammatory monocytes and CD11b+CD11c+ dendritic cells in a mouse model of EAE; reduces hepatic fibrosis and accumulation of inflammatory monocytes and neutrophils in the N-IF mouse model of liver fibrosis at 25 mg/kg per day; delays disease onset and decreases insulitis in NOD mice; reduces skin thickness, the number of myofibroblasts, and autoantibody production in the tight skin 1 mouse model of systemic sclerosis at 5 and 25 mg/kg