Learn More
Cayman Chemical Parishin C, Purity: ≥95%, CAS: 174972-80-6, 10 mg
SDP
Catalog No. 502934155
Encompass_Preferred
A phenolic glycoside; prevents Aβ42-induced inhibition of NMDAR currents in rat hippocampal neurons at 2 μM; prevents Aβ42-induced inhibition of LTP in rats at 20 mg/kg; a 5-HT1A receptor agonist (Ki = 1.54 nM; EC50 = 34 nM for [35S]GTPγS binding); reverses phencyclidine-induced increases in immobility time in the forced swim test, phencyclidine-induced decreases in interaction time in the social interaction test, and reverses phencyclidine-induced impairments in visual recognition of a novel object in mice
Catalog No. 50-293-4155 Supplier Cayman Chemical Supplier No. 2512710 MG
A phenolic glycoside; prevents Aβ42-induced inhibition of NMDAR currents in rat hippocampal neurons at 2 μM; prevents Aβ42-induced inhibition of LTP in rats at 20 mg/kg; a 5-HT1A receptor agonist (Ki = 1.54 nM; EC50 = 34 nM for [35S]GTPγS binding); reverses phencyclidine-induced increases in immobility time in the forced swim test, phencyclidine-induced decreases in interaction time in the social interaction test, and reverses phencyclidine-induced impairments in visual recognition of a novel object in mice