Learn More
Cayman Chemical PD 150606, Purity: ≥98%, CAS: 179528-45-1, 5 mg
SDP
Catalog No. 502955192
Encompass_Preferred
A selective, cell-permeable inhibitor of calpains (Ki = 0.21 μM for μ-calpain (calpain-1) and 0.37 μM for m-calpain (calpain-2))
Catalog No. 50-295-5192 Supplier Cayman Chemical Supplier No. 138595 MG
A selective, cell-permeable inhibitor of calpains (Ki = 0.21 μM for μ-calpain (calpain-1) and 0.37 μM for m-calpain (calpain-2))