Learn More
Cayman Chemical PD 404182, Purity: ≥98%, CAS: 72596-74-8, 10 mg
SDP
Catalog No. 502950909
Encompass_Preferred
An antiviral agent; inhibits SARS-CoV-2 Mpro activity (IC50 = 0.081 µM); prevents HSV-1 and HSV-2 infection in Vero cells at 0.2 µM; inhibits DDAH-1 and HDAC8 (IC50s = 9 and 0.011 µM for the human enzymes, respectively); inhibits bacterial KDO 8-P synthase (Ki = 0.026 µM); reduces endothelial tube formation and LPS-induced NO production in primary human dermal microvascular endothelial cells
Catalog No. 50-295-0909 Supplier Cayman Chemical Supplier No. 3439610 MG
An antiviral agent; inhibits SARS-CoV-2 Mpro activity (IC50 = 0.081 µM); prevents HSV-1 and HSV-2 infection in Vero cells at 0.2 µM; inhibits DDAH-1 and HDAC8 (IC50s = 9 and 0.011 µM for the human enzymes, respectively); inhibits bacterial KDO 8-P synthase (Ki = 0.026 µM); reduces endothelial tube formation and LPS-induced NO production in primary human dermal microvascular endothelial cells