Learn More
Cayman Chemical Pencycuron, Purity: ≥95%, CAS: 66063-05-6, 500 mg
SDP
Catalog No. 502912686
Encompass_Preferred
A phenylurea fungicide; active against the plant pathogenic fungi G. graminis, R. oryzae, and R. solani, but not P. intermedium or P. irregulare, at 10 µg/ml; reduces the incidence of R. solani-induced sheath blight on rice at 125, 250, and 500 ppm
Catalog No. 50-291-2686 Supplier Cayman Chemical Supplier No. 36127500 MG
A phenylurea fungicide; active against the plant pathogenic fungi G. graminis, R. oryzae, and R. solani, but not P. intermedium or P. irregulare, at 10 µg/ml; reduces the incidence of R. solani-induced sheath blight on rice at 125, 250, and 500 ppm