Learn More
Cayman Chemical ANTIBODY PEO-IAA 100mg
SDP
Catalog No. 502944329
Encompass_Preferred
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 UG/L in growth media
Catalog No. 50-294-4329 Supplier Cayman Chemical Supplier No. 40665100 MG
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 UG/L in growth media