Learn More
Cayman Chemical PEO-IAA, Purity: ≥98%, CAS: 6266-66-6, 1 g
SDP
Catalog No. 502943136
Encompass_Preferred
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 µg/L in growth media
Catalog No. 50-294-3136 Supplier Cayman Chemical Supplier No. 406651 G
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 µg/L in growth media