Learn More
Cayman Chemical PEO-IAA, Purity: ≥98%, CAS: 6266-66-6, 500 mg
SDP
Catalog No. 502944202
Encompass_Preferred
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 µg/L in growth media
Catalog No. 50-294-4202 Supplier Cayman Chemical Supplier No. 40665500 MG
An auxin antagonist; slows shoot growth and suppresses auxin-induced apical dominance in C. sativa seedlings at 10 µg/L in growth media