Learn More
Cayman Chemical PF-573228, Purity: ≥95%, CAS: 869288-64-2, 50 mg
SDP
Catalog No. 502973098
Encompass_Preferred
A selective FAK inhibitor (IC50s = 4 and 30-100 nM for a purified recombinant catalytic fragment of FAK and in cultured cells, respectively) that can inhibit chemotactic and haptotactic migration of cells as well as prevent focal adhesion turnover
Catalog No. 50-297-3098 Supplier Cayman Chemical Supplier No. 1492450 MG
A selective FAK inhibitor (IC50s = 4 and 30-100 nM for a purified recombinant catalytic fragment of FAK and in cultured cells, respectively) that can inhibit chemotactic and haptotactic migration of cells as well as prevent focal adhesion turnover