Learn More
Cayman Chemical PFK15, Purity: ≥95%, CAS: 4382-63-2, 5 mg
SDP
Catalog No. 502979557
Encompass_Preferred
A selective inhibitor of PFKFB3 (IC50 = 207 nM); rapidly induces apoptosis in transformed cells, suppresses glucose uptake and growth of Lewis lung carcinomas in syngeneic mice, and yields antitumor effects in human xenograft models of cancer in athymic mice
Catalog No. 50-297-9557 Supplier Cayman Chemical Supplier No. 176895 MG
A selective inhibitor of PFKFB3 (IC50 = 207 nM); rapidly induces apoptosis in transformed cells, suppresses glucose uptake and growth of Lewis lung carcinomas in syngeneic mice, and yields antitumor effects in human xenograft models of cancer in athymic mice