Learn More
Cayman Chemical PHA-408, Purity: ≥98%, CAS: 503555-55-3, 1 mg
SDP
Catalog No. 502944932
Encompass_Preferred
An IKKβ inhibitor (IC50 = 0.04 µM); selective for IKKβ over IKKα, Pim-1, Pim-2, Tie2, TrkA, and p70S6K (IC50s = 0.6-28.4 µM), as well as a panel of 26 kinases (IC50s = >30 µM); inhibits IL-1β-induced PGE2 production in synovial fibroblasts isolated from patients with rheumatoid arthritis (IC50 = ~0.6 µM); reduces serum LPS-induced TNF-α production in rats at 50 mg/kg; inhibits joint swelling and bone destruction in a rat model of streptococcal cell wall-induced arthritis at 10 mg/kg; inhibits LPS- or cigarette smoke-induced BALF neutrophil influx and reduces BALF levels of IL-6, TNF-α, and IL-1β in rats at 15 and 45 mg/kg
Catalog No. 50-294-4932 Supplier Cayman Chemical Supplier No. 412791 MG
An IKKβ inhibitor (IC50 = 0.04 µM); selective for IKKβ over IKKα, Pim-1, Pim-2, Tie2, TrkA, and p70S6K (IC50s = 0.6-28.4 µM), as well as a panel of 26 kinases (IC50s = >30 µM); inhibits IL-1β-induced PGE2 production in synovial fibroblasts isolated from patients with rheumatoid arthritis (IC50 = ~0.6 µM); reduces serum LPS-induced TNF-α production in rats at 50 mg/kg; inhibits joint swelling and bone destruction in a rat model of streptococcal cell wall-induced arthritis at 10 mg/kg; inhibits LPS- or cigarette smoke-induced BALF neutrophil influx and reduces BALF levels of IL-6, TNF-α, and IL-1β in rats at 15 and 45 mg/kg