Learn More
Cayman Chemical PHA-665752, Purity: ≥98%, CAS: 477575-56-7, 10 mg
SDP
Catalog No. 502972592
Encompass_Preferred
An active-site inhibitor of the catalytic activity of c-Met kinase (Ki = 4 nM; IC50 = 9 nM) that exhibits >50-fold selectivity for c-Met over a panel of tyrosine and serine-threonine kinases; inhibits c-Met phosphorylation, as well as cell proliferation and cell motility, of various tumor cells (IC50s = 18-50 nM)
Catalog No. 50-297-2592 Supplier Cayman Chemical Supplier No. 1470310 MG
An active-site inhibitor of the catalytic activity of c-Met kinase (Ki = 4 nM; IC50 = 9 nM) that exhibits >50-fold selectivity for c-Met over a panel of tyrosine and serine-threonine kinases; inhibits c-Met phosphorylation, as well as cell proliferation and cell motility, of various tumor cells (IC50s = 18-50 nM)