Learn More
Cayman Chemical PHA-848125, Purity: ≥98%, CAS: 802539-81-7, 10 mg
SDP
Catalog No. 502986262
Encompass_Preferred
An ATP-competitive inhibitor of Cdks that most potently inhibits Cdk2/cyclin A (IC50 = 45 nM); more than 3-fold less potent at Cdks 1, 3, 4, 5, and 7
Catalog No. 50-298-6262 Supplier Cayman Chemical Supplier No. 2147410 MG
An ATP-competitive inhibitor of Cdks that most potently inhibits Cdk2/cyclin A (IC50 = 45 nM); more than 3-fold less potent at Cdks 1, 3, 4, 5, and 7