Learn More
Cayman Chemical Phleomycin, Purity: ≥95% (mixture of phleomycins), CAS: 11006-33-0, 50 mg
SDP
Catalog No. 502969500
Encompass_Preferred
A glycopeptide antibiotic with diverse biological activities; induces chromosomal breaks in S. cerevisiae at 1 to 2 µM; reduces proliferation, DNA synthesis, and the percentage of cells entering mitosis in HeLa cells at 5 or 10 µg/ml; increases survival time in combination with LS34 in an Ehrlich mouse ascites tumor model at 1 mg/kg per day; has been used as an antibiotic selection agent for successfully transformed cells in genetic engineering experiments
Catalog No. 50-296-9500 Supplier Cayman Chemical Supplier No. 1554950 MG
A glycopeptide antibiotic with diverse biological activities; induces chromosomal breaks in S. cerevisiae at 1 to 2 µM; reduces proliferation, DNA synthesis, and the percentage of cells entering mitosis in HeLa cells at 5 or 10 µg/ml; increases survival time in combination with LS34 in an Ehrlich mouse ascites tumor model at 1 mg/kg per day; has been used as an antibiotic selection agent for successfully transformed cells in genetic engineering experiments