Learn More
Cayman Chemical Physcion, Purity: ≥98%, CAS: 521-61-9, 10 mg
SDP
Catalog No. 502980694
Encompass_Preferred
A natural anthraquinone that has been reported to have laxative and anti-inflammatory actions, inhibit kinases and tyrosinase, and have cytotoxicity against cancer cells; inhibits 6-PGD (IC50 = 38.5 µM), decreasing cell proliferation and tumor growth in mice
Catalog No. 50-298-0694 Supplier Cayman Chemical Supplier No. 1986310 MG
A natural anthraquinone that has been reported to have laxative and anti-inflammatory actions, inhibit kinases and tyrosinase, and have cytotoxicity against cancer cells; inhibits 6-PGD (IC50 = 38.5 µM), decreasing cell proliferation and tumor growth in mice