Learn More
Cayman Chemical PI-828, Purity: ≥98%, CAS: 942289-87-4, 5 mg
SDP
Catalog No. 502986451
Encompass_Preferred
A potent inhibitor of PI3K (IC50s = 9.8, 183, 227, and 1,967 nM for p110β, p110α, p110δ, and p110γ, respectively); has been immobilized on solid surface; interacts with non-PI3K cellular proteins such as mTOR, ALDH1, BRD4, CKIIα, and GSK-3β.
Catalog No. 50-298-6451 Supplier Cayman Chemical Supplier No. 218115 MG
A potent inhibitor of PI3K (IC50s = 9.8, 183, 227, and 1,967 nM for p110β, p110α, p110δ, and p110γ, respectively); has been immobilized on solid surface; interacts with non-PI3K cellular proteins such as mTOR, ALDH1, BRD4, CKIIα, and GSK-3β.