Learn More
Cayman Chemical Picamilon, Purity: ≥98%, CAS: 34562-97-5, 500 mg
SDP
Catalog No. 502991198
Encompass_Preferred
A GABA derivative; increases parietal cortical blood flow in rats and rabbits at 50 mg/kg, i.p. and 10 mg/kg, i.v., respectively; increases latency to enter the dark compartment in the passive avoidance conditioned reflex test in rats at 10 mg/kg compared with control animals
Catalog No. 50-299-1198 Supplier Cayman Chemical Supplier No. 29397500 MG
A GABA derivative; increases parietal cortical blood flow in rats and rabbits at 50 mg/kg, i.p. and 10 mg/kg, i.v., respectively; increases latency to enter the dark compartment in the passive avoidance conditioned reflex test in rats at 10 mg/kg compared with control animals