Learn More
Cayman Chemical Picrotin, Purity: ≥98%, CAS: 21416-53-5, 100 mg
SDP
Catalog No. 502986361
Encompass_Preferred
A natural antagonist of GlyRs (IC50s = 57 and 117 µM for α1 and α2 homodimeric GlyRs, respectively); does not inhibit GABAA or GABAC receptors
Catalog No. 50-298-6361 Supplier Cayman Chemical Supplier No. 21870100 MG
A natural antagonist of GlyRs (IC50s = 57 and 117 µM for α1 and α2 homodimeric GlyRs, respectively); does not inhibit GABAA or GABAC receptors