Learn More
Cayman Chemical Pidotimod, Purity: ≥98%, CAS: 121808-62-6, 5 g
SDP
Catalog No. 502968335
Encompass_Preferred
A synthetic dipeptide with immunomodulatory properties on both innate and adaptive immune responses in in vitro studies
Catalog No. 50-296-8335 Supplier Cayman Chemical Supplier No. 187255 G
A synthetic dipeptide with immunomodulatory properties on both innate and adaptive immune responses in in vitro studies