Learn More
Cayman Chemical PIK-293, Purity: ≥98%, CAS: 900185-01-5, 1 mg
SDP
Catalog No. 502985450
Encompass_Preferred
A selective inhibitor of the PI3K catalytic subunit p110δ (IC50s = 0.24 µM)
Catalog No. 50-298-5450 Supplier Cayman Chemical Supplier No. 212831 MG
A selective inhibitor of the PI3K catalytic subunit p110δ (IC50s = 0.24 µM)