Learn More
Cayman Chemical Pindolol, Purity: ≥99%, CAS: 13523-86-9, 500 mg
SDP
Catalog No. 502994646
Encompass_Preferred
A β1- and β2-AR antagonist (Kis = 2.6 and 4.8 nM, respectively); partial agonist of the β3-AR; antagonist of the 5-HT1A receptor (Ki = 81.1 nM for inhibition of 5-HT-stimulated GTPγS binding); inhibits isoproterenol-induced tachycardia in anesthetized cats (ED50 = 1.8 µg/kg); decreases mean blood pressure in conscious spontaneously hypertensive rats at 30 mg/kg per day
Catalog No. 50-299-4646 Supplier Cayman Chemical Supplier No. 21445500 MG
A β1- and β2-AR antagonist (Kis = 2.6 and 4.8 nM, respectively); partial agonist of the β3-AR; antagonist of the 5-HT1A receptor (Ki = 81.1 nM for inhibition of 5-HT-stimulated GTPγS binding); inhibits isoproterenol-induced tachycardia in anesthetized cats (ED50 = 1.8 µg/kg); decreases mean blood pressure in conscious spontaneously hypertensive rats at 30 mg/kg per day