Learn More
Cayman Chemical Pirodavir, Purity: ≥98%, CAS: 124436-59-5, 25 mg
SDP
Catalog No. 502994907
Encompass_Preferred
A broad-spectrum antipicornaviral agent; active against 80 typed (MICs = 0.001-14.4 µg/ml) and one untyped rhinovirus strain (MIC = 0.002 µg/ml); active against serotypes of coxsackievirus, poliovirus, and echovirus enteroviruses (MICs = 0.018-8.5, 0.025-0.27, and 0.019-1.3 µg/ml, respectively)
Catalog No. 50-299-4907 Supplier Cayman Chemical Supplier No. 2292225 MG
A broad-spectrum antipicornaviral agent; active against 80 typed (MICs = 0.001-14.4 µg/ml) and one untyped rhinovirus strain (MIC = 0.002 µg/ml); active against serotypes of coxsackievirus, poliovirus, and echovirus enteroviruses (MICs = 0.018-8.5, 0.025-0.27, and 0.019-1.3 µg/ml, respectively)