Learn More
Cayman Chemical Plerixafor, Purity: ≥95%, CAS: 110078-46-1, 1 g
SDP
Catalog No. 502913443
Encompass_Preferred
An antagonist of CXCR4 (IC50s = 0.02-0.13 µg/ml); selective for CXCR4 over CXCR1-3 and CXCR5-9 (IC50s = >25 µg/ml); decreases infectious virus content in the supernatant of Jurkat cells chronically infected with HIV-1(IIIB) (EC50 = ~0.02 µg/ml); mobilizes murine and human hematopoietic stem and murine long-term repopulating cells for transplantation; increases T cell trafficking in mouse blood, spleen, and central nervous system; decreases the number of 4T1 murine mammary carcinoma cells in the lung in a mouse model of lung metastasis at 1.25 mg/kg twice per day
Catalog No. 50-291-3443 Supplier Cayman Chemical Supplier No. 355791 G
An antagonist of CXCR4 (IC50s = 0.02-0.13 µg/ml); selective for CXCR4 over CXCR1-3 and CXCR5-9 (IC50s = >25 µg/ml); decreases infectious virus content in the supernatant of Jurkat cells chronically infected with HIV-1(IIIB) (EC50 = ~0.02 µg/ml); mobilizes murine and human hematopoietic stem and murine long-term repopulating cells for transplantation; increases T cell trafficking in mouse blood, spleen, and central nervous system; decreases the number of 4T1 murine mammary carcinoma cells in the lung in a mouse model of lung metastasis at 1.25 mg/kg twice per day