Learn More
Cayman Chemical PNU 120596, Purity: ≥98%, CAS: 501925-31-1, 25 mg
SDP
Catalog No. 502954196
Encompass_Preferred
A positive allosteric modulator of the α7 neuronal nAChR, increasing agonist-evoked calcium flux (EC50 = 216 nM); increases the frequency of ACh-evoked GABAergic postsynaptic currents in rat hippocampal slices and improves the auditory gating deficit caused by amphetamine in rats
Catalog No. 50-295-4196 Supplier Cayman Chemical Supplier No. 1430425 MG
A positive allosteric modulator of the α7 neuronal nAChR, increasing agonist-evoked calcium flux (EC50 = 216 nM); increases the frequency of ACh-evoked GABAergic postsynaptic currents in rat hippocampal slices and improves the auditory gating deficit caused by amphetamine in rats