Learn More
Cayman Chemical PQ401, Purity: ≥98%, CAS: 196868-63-0, 10 mg
SDP
Catalog No. 502983677
Encompass_Preferred
An inhibitor of IGF-1R; inhibits autophosphorylation of the recombinant IGF-1R kinase domain (IC50 = 50 = 12 μM); reduces cell viability of U87MG glioma cells (IC50 = 5 μM) and induces apoptosis and inhibits cell migration in a concentration-dependent manner; inhibits IGF-1-stimulated proliferation of MCF-7 cells in vitro (IC50 = 6 μM) and reduces tumor growth in an MCNeuA model of murine breast cancer at 50 and 100 mg/kg; has antimalarial activity against P. falciparum strain 3D7 (EC50 = 53 nM)
Catalog No. 50-298-3677 Supplier Cayman Chemical Supplier No. 2097910 MG
An inhibitor of IGF-1R; inhibits autophosphorylation of the recombinant IGF-1R kinase domain (IC50 = 50 = 12 μM); reduces cell viability of U87MG glioma cells (IC50 = 5 μM) and induces apoptosis and inhibits cell migration in a concentration-dependent manner; inhibits IGF-1-stimulated proliferation of MCF-7 cells in vitro (IC50 = 6 μM) and reduces tumor growth in an MCNeuA model of murine breast cancer at 50 and 100 mg/kg; has antimalarial activity against P. falciparum strain 3D7 (EC50 = 53 nM)