Learn More
Cayman Chemical PRT060318, Purity: ≥98%, CAS: 1194961-19-7, 5 mg
SDP
Catalog No. 502986467
Encompass_Preferred
A potent and selective Syk inhibitor (IC50 = 4 nM); inhibits 92% of Syk activity, while other kinases retain >70% activity, at a concentration of 50 nM in a panel of 270 kinases; inhibits convulxin-induced aggregation of human platelet-rich plasma (IC50 = 2.5 μM); prevents thrombosis in a transgenic mouse model of heparin-induced thrombocytopenia; induces CLL B cell apoptosis and inhibits the secretion of chemokines CCL3, CCL4, and CXCL13; inhibits CLL B cell chemotaxis and pseudoemperipolesis
Catalog No. 50-298-6467 Supplier Cayman Chemical Supplier No. 224765 MG
A potent and selective Syk inhibitor (IC50 = 4 nM); inhibits 92% of Syk activity, while other kinases retain >70% activity, at a concentration of 50 nM in a panel of 270 kinases; inhibits convulxin-induced aggregation of human platelet-rich plasma (IC50 = 2.5 μM); prevents thrombosis in a transgenic mouse model of heparin-induced thrombocytopenia; induces CLL B cell apoptosis and inhibits the secretion of chemokines CCL3, CCL4, and CXCL13; inhibits CLL B cell chemotaxis and pseudoemperipolesis