Learn More
Cayman Chemical PSC 833, Purity: ≥95%, CAS: 121584-18-7, 1 mg
SDP
Catalog No. 502983673
Encompass_Preferred
A multidrug-resistance modulator; restores sensitivity of MDR-P388 murine leukemia cells to various cytostatic agents from 26-70 nM; inhibits basal-to-apical transport and increases apical-to-basal transport of [14C]docetaxel in LLC-GA5-COLO150 cells that overexpress human P-gp; increases survival time in MDR-P388 tumor-bearing mice and in an MDR-L1210 leukemia mouse xenograft model when administered in combination with doxorubicin; prolongs the anti-hyperalgesic effects of intraperitoneally administered pregabalin in a mouse model of cold stress-induced central pain
Catalog No. 50-298-3673 Supplier Cayman Chemical Supplier No. 203911 MG
A multidrug-resistance modulator; restores sensitivity of MDR-P388 murine leukemia cells to various cytostatic agents from 26-70 nM; inhibits basal-to-apical transport and increases apical-to-basal transport of [14C]docetaxel in LLC-GA5-COLO150 cells that overexpress human P-gp; increases survival time in MDR-P388 tumor-bearing mice and in an MDR-L1210 leukemia mouse xenograft model when administered in combination with doxorubicin; prolongs the anti-hyperalgesic effects of intraperitoneally administered pregabalin in a mouse model of cold stress-induced central pain