Learn More
Cayman Chemical PSI-7977, Purity: ≥98%, CAS: 1190307-88-0, 10 mg
SDP
Catalog No. 502973142
Encompass_Preferred
A prodrug form of PSI-7411; inhibits HCV replication in an HCV replicon assay (EC50 = 0.092 µM); inhibits SARS-CoV-2 replication in iPSC-derived human brain cortical organoids at 20 µM
Catalog No. 50-297-3142 Supplier Cayman Chemical Supplier No. 1540210 MG
A prodrug form of PSI-7411; inhibits HCV replication in an HCV replicon assay (EC50 = 0.092 µM); inhibits SARS-CoV-2 replication in iPSC-derived human brain cortical organoids at 20 µM