Learn More
Abcam Antibody Purification Kit (Protein G), 3 Tests
Catalog No. 502584237
Encompass_Preferred
Commercially available antibodies often contain substances (e.g. BSA, glycine, tris, azide) that interfere in labeling reactions with enzymes or fluorophores. ab128747 quickly removes these contaminants.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4237 Supplier Abcam Supplier No. AB1287473TEST
Commercially available antibodies often contain substances (e.g. BSA, glycine, tris, azide) that interfere in labeling reactions with enzymes or fluorophores. ab128747 quickly removes these contaminants.
The product is subject to the following: Abcam Restricted Use Statement