Learn More
Cayman Chemical PX 1, Purity: ≥98%, CAS: 2221100-71-4, 10 mg
SDP
Catalog No. 502970745
Encompass_Preferred
A derivative of AM2201 with a PINACA-like upper chain where the isopropyl group has been replaced with a benzyl group; intended for research and forensic applications
Catalog No. 50-297-0745 Supplier Cayman Chemical Supplier No. 1620110 MG
A derivative of AM2201 with a PINACA-like upper chain where the isopropyl group has been replaced with a benzyl group; intended for research and forensic applications