Learn More
Cayman Chemical Pyrogallol, Purity: ≥98%, CAS: 87-66-1, 250 g
SDP
Catalog No. 502980756
Encompass_Preferred
A natural oxidant that can generate O2- in alkaline solutions through autoxidation to a semiquinone radical, which in turn can react with O2- in an acidic environment to produce a quinone and H2O2; used in superoxide dismutase and antioxidant assays, as well as in biological systems
Catalog No. 50-298-0756 Supplier Cayman Chemical Supplier No. 20347250 G
A natural oxidant that can generate O2- in alkaline solutions through autoxidation to a semiquinone radical, which in turn can react with O2- in an acidic environment to produce a quinone and H2O2; used in superoxide dismutase and antioxidant assays, as well as in biological systems