Learn More
Cayman Chemical pythiDC, Purity: ≥98%, CAS: 1821370-71-1, 10 mg
SDP
Catalog No. 502919415
Encompass_Preferred
A CP4H1 inhibitor (IC50 = 4 µM); selective for CP4H1 over HIF-PH2 at 10 µM
Catalog No. 50-291-9415 Supplier Cayman Chemical Supplier No. 3739210 MG
A CP4H1 inhibitor (IC50 = 4 µM); selective for CP4H1 over HIF-PH2 at 10 µM