Learn More
Cayman Chemical Quinine, Purity: ≥95%, CAS: 130-95-0, 10 g
SDP
Catalog No. 502994521
Encompass_Preferred
An antimalarial agent (MICs = 10-500 nM for P. falciparum); inhibits hemozoin formation in purified trophozoites, leading to an increase in free heme; reduces mouse P. berghei parasite load in the blood with an MED of 150 mg/kg
Catalog No. 50-299-4521 Supplier Cayman Chemical Supplier No. 2395810 G
An antimalarial agent (MICs = 10-500 nM for P. falciparum); inhibits hemozoin formation in purified trophozoites, leading to an increase in free heme; reduces mouse P. berghei parasite load in the blood with an MED of 150 mg/kg