Learn More
Cayman Chemical R 1530, Purity: ≥98%, CAS: 882531-87-5, 5 mg
SDP
Catalog No. 502969260
Encompass_Preferred
A multi-kinase inhibitor that targets over 20 kinases, including angiogenesis-related receptor tyrosine kinases (Kds = 61, 88, and 15 nM for FGFR1, PDGFRβ, and VEGFR2, respectively); also inhibits FLT1, KIT, PLK4, and RET with Kd values of 9, 26, 11, and 22 nM, respectively
Catalog No. 50-296-9260 Supplier Cayman Chemical Supplier No. 152555 MG
A multi-kinase inhibitor that targets over 20 kinases, including angiogenesis-related receptor tyrosine kinases (Kds = 61, 88, and 15 nM for FGFR1, PDGFRβ, and VEGFR2, respectively); also inhibits FLT1, KIT, PLK4, and RET with Kd values of 9, 26, 11, and 22 nM, respectively