Learn More
Cayman Chemical ANTIBODY R-7050 10mg
SDP
Catalog No. 502980296
Encompass_Preferred
A cell-permeable antagonist of signaling through the TNF-α receptor complex (EC50 = 0.631 µM), blocking TNF-α-induced binding of TNF-α receptor I with TRADD and RIP1
Catalog No. 50-298-0296 Supplier Cayman Chemical Supplier No. 1687010 MG
A cell-permeable antagonist of signaling through the TNF-α receptor complex (EC50 = 0.631 µM), blocking TNF-α-induced binding of TNF-α receptor I with TRADD and RIP1