Learn More
Cayman Chemical R-848, Purity: ≥98%, CAS: 144875-48-9, 10 mg
SDP
Catalog No. 502972278
Encompass_Preferred
An immune response modifier that acts as a ligand of TLR7 and 8; demonstrates potent anti-tumor and anti-viral properties and has been used to study the innate immune response to viral infections
Catalog No. 50-297-2278 Supplier Cayman Chemical Supplier No. 1480610 MG
An immune response modifier that acts as a ligand of TLR7 and 8; demonstrates potent anti-tumor and anti-viral properties and has been used to study the innate immune response to viral infections