Learn More
Cayman Chemical R428, Purity: ≥98%, CAS: 1037624-75-1, 25 mg
SDP
Catalog No. 502986127
Encompass_Preferred
An inhibitor of Axl kinase (EC50 = 14 nM); orally bioavailable, blocks angiogenesis in corneal micropocket and tumor models, and reduces metastatic burden while extending survival in mouse models of breast cancer metastasis
Catalog No. 50-298-6127 Supplier Cayman Chemical Supplier No. 2152325 MG
An inhibitor of Axl kinase (EC50 = 14 nM); orally bioavailable, blocks angiogenesis in corneal micropocket and tumor models, and reduces metastatic burden while extending survival in mouse models of breast cancer metastasis