Learn More
Cayman Chemical RA-9, Purity: ≥98%, CAS: 919091-63-7, 5 mg
SDP
Catalog No. 502978586
Encompass_Preferred
An inhibitor of deubiquitinating enzymes; inhibits UCH-L1, UCH-L3, and USP8 in a cell-free assay at 10 µM, as well as USP5 in a reporter assay at the same concentration; inhibits the 26S proteasome, but not the 20S proteasome, in a reporter assay using HeLa cancer cells at 10 µM; increases the levels of polyubiquitinated proteins in HeLa and TOV21G1 cancer cells at 5 µM; induces cell cycle arrest at the S and G2/M phases in HeLa cells at 10 µM; decreases the viability of nine cancer cell lines (IC50s= 1.64-12.49 µM); increases survival and decreases tumor and ascitic fluid mass in an ES-2 ovarian cancer mouse xenograft model at 5 mg/kg every other day
Catalog No. 50-297-8586 Supplier Cayman Chemical Supplier No. 408385 MG
An inhibitor of deubiquitinating enzymes; inhibits UCH-L1, UCH-L3, and USP8 in a cell-free assay at 10 µM, as well as USP5 in a reporter assay at the same concentration; inhibits the 26S proteasome, but not the 20S proteasome, in a reporter assay using HeLa cancer cells at 10 µM; increases the levels of polyubiquitinated proteins in HeLa and TOV21G1 cancer cells at 5 µM; induces cell cycle arrest at the S and G2/M phases in HeLa cells at 10 µM; decreases the viability of nine cancer cell lines (IC50s= 1.64-12.49 µM); increases survival and decreases tumor and ascitic fluid mass in an ES-2 ovarian cancer mouse xenograft model at 5 mg/kg every other day