Learn More
Cayman Chemical ANTIBODY RAD140 500ug
SDP
Catalog No. 502967822
Encompass_Preferred
An orally bioavailable SARM that binds to the androgen receptor with a Ki value of 7 nM; stimulates muscle weight increase without affecting prostate or seminal vesicle in castrated male rats
Catalog No. 50-296-7822 Supplier Cayman Chemical Supplier No. 18773500 UG
An orally bioavailable SARM that binds to the androgen receptor with a Ki value of 7 nM; stimulates muscle weight increase without affecting prostate or seminal vesicle in castrated male rats