Learn More
Cayman Chemical ANTIBODY RadIcIcol 500ug
SDP
Catalog No. 502953232
Encompass_Preferred
An inhibitor of Hsp90 (IC50 = erb82, and mutant p53; binds to and inhibits DNA topoisomerase type II proteins and GRP94
Catalog No. 50-295-3232 Supplier Cayman Chemical Supplier No. 13089500 UG
An inhibitor of Hsp90 (IC50 = erb82, and mutant p53; binds to and inhibits DNA topoisomerase type II proteins and GRP94