Learn More
Cayman Chemical Reparixin, Purity: ≥95%, CAS: 266359-83-5, 10 mg
SDP
Catalog No. 502980473
Encompass_Preferred
An allosteric antagonist of CXCR1 and CXCR2 (IC50s = 1 and 100 nM, respectively); reduces ischemia and reperfusion-induced hepatic PMN recruitment, hepatic MPO levels, and serum ALT levels in a rat model of hepatic ischemia at 3, 15, and 30 mg/kg; decreases systolic blood pressure in spontaneously hypertensive rats at 5 mg/kg; reduces tumor volume and the number of intratumor cancer stem cells in a SUM159 mouse xenograft model when administered in combination with docetaxel
Catalog No. 50-298-0473 Supplier Cayman Chemical Supplier No. 1735010 MG
An allosteric antagonist of CXCR1 and CXCR2 (IC50s = 1 and 100 nM, respectively); reduces ischemia and reperfusion-induced hepatic PMN recruitment, hepatic MPO levels, and serum ALT levels in a rat model of hepatic ischemia at 3, 15, and 30 mg/kg; decreases systolic blood pressure in spontaneously hypertensive rats at 5 mg/kg; reduces tumor volume and the number of intratumor cancer stem cells in a SUM159 mouse xenograft model when administered in combination with docetaxel