Learn More
BioVendor, LLC Antibody Resistin Hu Pabantib BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD181016220
Encompass
TypePolyclonal AntibodyApplicationsWestern blotting, Immunohistochemistry Source of AntigenE. coliHostsRabbitPreparationThe antibody was raised in rabbits by immunization with the recombinant Human Resistin.Species ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human Resistin.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)FormulationThe antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.ReconstitutionAdd 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.+C41
Catalog No. 50-894-157